


(S,S,S)-Triglycidyl isocyanurate ≥98.0% (by total nitrogen basis)
Catalog # TCIAI0581-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Melting point:105.0 to 108.0 deg-C
- Specific rotation [a]20/D:-20.0 to -24.0 deg (C=0.4, H2O)
Specification Test Results
About this item
:
UN:2811
ADR:6.1,III
:GHS05
:GHS06
:GHS08
CAS Number:240408-81-5
Formula:C₁₂H₁₅N₃O₆
MDL Number:MFCD03844799