


(R,R,R)-Triglycidyl isocyanurate ≥98.0% (by GC)
TCIAI0580-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:104.0 to 108.0 deg-C
- Specific rotation [a]20/D:+9.0 to +12.0 deg (C=1, CH3CN (50%))
:
:2811
:6.1,III
:GHS05
:GHS06
:GHS08
:240408-78-0
:C₁₂H₁₅N₃O₆
:MFCD03844798