Order Entry
Netherlands
ContactUsLinkComponent
Bathophenanthrolinedisulphonic acid disodium salt hydrate for determination of ferrous ion
Bathophenanthrolinedisulphonic acid disodium salt hydrate for determination of ferrous ion
Catalog # TCIAB0989-1G
Supplier:  TCI
CAS Number:  
undefined
Bathophenanthrolinedisulphonic acid disodium salt hydrate for determination of ferrous ion
Catalog # TCIAB0989-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Suitability for iron-analysis:
    min. 0.50 (535 nm)

Specification Test Results

About this item

 :
 :536.48 g/mol (anhy)
EINECS:258-740-7
CAS Number:53744-42-6
Formula:C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MDL Number:MFCD00038854