
1-Tritylimidazole-4-carbaldehyde ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:33016-47-6
:
Formula:C₂₃H₁₈N₂O
MDL Number:MFCD02179554
:338.41 g/mol
Melting Pt:186°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together