N(α)-Boc-D-tryptophan ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:5241-64-5
:
Formula:C₁₆H₂₀N₂O₄
MDL Number:MFCD00037944
:304.3 g/mol
EINECS:226-042-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:+15.0 to +24.0 deg (C=2, DMF)
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together