
(3-Aminopropyl)diethoxymethylsilane ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3179-76-8
:
Formula:C₈H₂₁NO₂Si
MDL Number:MFCD00039785
:191.35 g/mol
Boiling Pt:88°C
Flash Pt:85°C
EINECS:221-660-8
UN:3267
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together