Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Nipecotamide 95%

Nipecotamide 95%

Irritant
Nipecotamide 95%
Certificates
Nipecotamide 95%

2 Options Available Below

Product Details & Documents
Synonyms:  Piperidine-3-carboxamide
  • CAS Number:4138-26-5
  • Formula:C₆H₁₂N₂O
  • MDL Number:MFCD00005993
  • Molecular Weight:128.1700 g/mol
  • Melting Pt:102…108 °C
  • Storage Temperature:Ambient
  • EINECS:223-962-5

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    102 to 108 °C
  • Titration with HClO4:
    ≥94 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 96,60 €
    96,60 €
    1 * 5 g
       96,60 €

  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 508,00 €
    508,00 €
    1 * 50 g
       508,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.