
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride ≥99.0% (by titrimetric analysis)
Catalog # TCIAN0542-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:246.738 g/mol
Merck Index:14,06368
Melting Pt:258°C
EINECS:208-989-2
UN:2811
ADR:6.1,II
:GHS06
CAS Number:550-99-2
Formula:C₁₄H₁₅ClN₂
MDL Number:MFCD00012554