
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:550-99-2
:
Formula:C₁₄H₁₅ClN₂
MDL Number:MFCD00012554
:246.738 g/mol
Merck Index:14,6368
Melting Pt:257…261 °C
Storage Temperature:Ambient
EINECS:208-989-2
ADR:6.1,II
:GHS06
Frequently Bought Together