N(α)-Fmoc-L-tryptophan ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Fmoc-Trp-OH
CAS Number:35737-15-6
:
Formula:C₂₆H₂₂N₂O₄
MDL Number:MFCD00037126
:426.5 g/mol
EINECS:252-706-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:185.0 to 188.0 deg-C
- Specific rotation [a]20/D:-27.0 to -30.0 deg (C=1, DMF)
Specification Test Results
Frequently Bought Together