Organic reference standard, 2-Hydroxydimetridazole 100 µg/ml in acetone, Dr Ehrenstorfer
EHERXA12772050AC
:
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:936-05-0
- Description:2-Hydroxydimetridazole 100 µg/ml in acetone
- Formula:C₅H₇N₃O₃
- Description:Dimetridazole-2-hydroxy