Dimetridazole-[1-methyl-D3] 100 µg/ml in acetone, Dr Ehrenstorfer
Catalog # EHERXA12772010AC
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:64678-69-9
- Description:Dimetridazole-[1-methyl-D3] 100 µg/ml in acetone
- Formula:C₅₂H₃H₄N₃O₂
- Description:Dimetridazole D3