2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86404-63-9
:
Formula:C₁₀H₇F₂N₃O
MDL Number:MFCD02093825
:223.18 g/mol
Melting Pt:103…107 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together