
1-Methylimidazole-2-carbaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13750-81-7
:
Formula:C₅H₆N₂O
MDL Number:MFCD01321308
:110.12 g/mol
Boiling Pt:70…74 °C (1 mmHg)
Melting Pt:36…39 °C
Flash Pt:102 °C (216 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together