Order Entry
Netherlands
ContactUsLinkComponent
 
 
1-Methylimidazole-2-carbaldehyde 97%
 
undefined
 
1-Methylimidazole-2-carbaldehyde 97%

 

  • Appearance:
    White to yellow to orange Crystals or needles or crystalline mass and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    35 to 39 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together