1,3-Dimethoxy-2-(methylthio)benzene
Catalog # MAYBCD11814.50
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
CAS Number:33617-67-3
Formula:C₉H₁₂O₂S
MDL Number:MFCD00010334