Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole ≥90.0% (by titrimetric analysis)
Flammable
Irritant
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole ≥90.0% (by titrimetric analysis)
Catalog # TCIAH0896-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole ≥90.0% (by titrimetric analysis)
Catalog # TCIAH0896-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 90.0 %

Specification Test Results

About this item

 :
 :194.21 g/mol
EINECS:257-909-2
UN:1325
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:52431-78-4
Formula:C₇H₆N₄OS
MDL Number:MFCD00132898