Order Entry
Netherlands
ContactUsLinkComponent
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Irritant
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3928-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3928-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :457.35 g/mol
Melting Pt:176°C
 :GHS07
CAS Number:59159-39-6
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00075527