Order Entry
Netherlands
ContactUsLinkComponent
Lead(II) oxide (Litharge), granular gold-, silver-free
  029-930-11
 :  
Lead(II) oxide (Litharge), granular gold-, silver-free
  029-930-11
 :  
  • Pk:
    25 kg
  • Seq:
    4283
  • Appearance granularity:
    Granular
  • Appearance colour:
    Yellow to orange
  • Gold (Au):
    ∼ 1 ppb
  • Silver (Ag):
    max 0.16 ppm
 :
 :13,05431
 :1470 °C (1013 hPa)
 :888 °C
 :9.5 g/cm³ (20 °C)
 :215-267-0
 :2291
 :6.1,III
 :223.21 g/mol
 :GHS08
 :GHS07
 :GHS09
 :1317-36-8
 :PbO
 :MFCD00011164