Order Entry
Netherlands
ContactUsLinkComponent
HPLC columns, HyPURITY™ ADVANCE
HPLC columns, HyPURITY™ ADVANCE
Certificates
HPLC columns, HyPURITY™ ADVANCE
Specifications
  • Catalog No:
  • 555-7026
  • 555-7029
  • 555-7032
  • 555-7023
  • 555-7024
  • Description:
  • Particle size: 5 µm
  • Particle size: 5 µm
  • Particle size: 5 µm
  • Particle size: 5 µm
  • Particle size: 5 µm
  • Carbon load:
  • 9%
  • 9%
  • 9%
  • 9%
  • 9%
  • Surface area:
  • 200 m²/g
  • 200 m²/g
  • 200 m²/g
  • 200 m²/g
  • 200 m²/g
  • USP code:
  • L7
  • L7
  • L7
  • L7
  • L7