Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Dispenser tips, ritips®
Dispenser tips, ritips®
Marketsource
Supplier:   BIOZYM
Dispenser tips, ritips®
Supplier:  BIOZYM
Marketsource
Certificates
Dispenser tips, ritips®
16 Options Available Below
Product Details & Documents
These dispenser tips are compatible with Ritter ripette®, Eppendorf Multipette® 4780 and many other dispensing systems.
Bioclean® tips are sterile and free from DNase, RNase and pyrogens.
Delivery information: The 25 ml and 50 ml tips are supplied with adapter.
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    For use with
    Packed
    Availability
    Price
  • Standard
    0,05 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    0,05 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    0,5 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    0,5 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    1,25 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    1,25 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    2,5 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    2,5 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    5,0 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    5,0 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    12,5 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    12,5 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    100/pack
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    25 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    25/pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    25 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    25/pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
    Specifications:
    Marketsource
    Marketsource
  • Standard
    50 ml
    -
    Ritter ripette®, Eppendorf® Multipette 4780
    25/pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
    Specifications:
    Marketsource
    Marketsource
  • BioClean®
    50 ml
    +
    Ritter ripette®, Eppendorf® Multipette 4780
    25/pack
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST
    Specifications:
    Marketsource
    Marketsource