Order Entry
Netherlands
ContactUsLinkComponent
1,2,4-Trimethylcyclohexane (mixture of stereoisomers) ≥96.0% (by GC)
Flammable
Health
1,2,4-Trimethylcyclohexane (mixture of stereoisomers) ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Health
1,2,4-Trimethylcyclohexane (mixture of stereoisomers) ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 % (total of isomer)

Specification Test Results

Frequently Bought Together