
2-Bromo-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3749-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:283.96 g/mol
:GHS07
CAS Number:452972-12-2
Formula:C₁₁H₁₅BBrNO₂
MDL Number:MFCD06798246