

Propagyl bromide ≥97.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:106-96-7
:
Formula:C₃H₃Br
MDL Number:MFCD00000241
:118.96 g/mol
Boiling Pt:90°C
Melting Pt:–43 °C
Density:1,552 g/cm³ (20 °C)
Flash Pt:10°C
UN:2345
ADR:3,II
:Stabiised with magnesium oxide (MgO)
EINECS:203-447-1
:GHS02
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together