4,4'-Diacetoxybiphenyl ≥99.0% (by GC)
Catalog # TCIAD2266-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:162.0 to 166.0 deg-C
Specification Test Results
About this item
:
:270.28 g/mol
CAS Number:32604-29-8
Formula:C₁₆H₁₄O₄
MDL Number:MFCD00035973