1,3-Dibenzyloxybenzene ≥96.0% (by GC)
Catalog # TCIAB2129-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:71.0 to 76.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:290.3557 g/mol
CAS Number:3769-42-4
Formula:C₂₀H₁₈O₂
MDL Number:MFCD00690156