
4-(4-Cyanophenyl)morpholine ≥98.0% (by GC)
Catalog # TCIAC2008-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:83.0 to 87.0 deg-C
Specification Test Results
About this item
:
:188.23 g/mol
UN:3439
ADR:6.1,III
:GHS06
CAS Number:10282-31-2
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD00100115