Order Entry
Netherlands
ContactUsLinkComponent
5-(4-Pyridyl)-1H-tetrazole ≥98%
5-(4-Pyridyl)-1H-tetrazole ≥98%
Catalog # B20741.03
CAS Number:  
5-(4-Pyridyl)-1H-tetrazole ≥98%
Catalog # B20741.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :147.14 g/mol
Storage Temperature:Ambient
CAS Number:14389-12-9
Formula:C₆H₅N₅
MDL Number:MFCD00068115