3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) 5% in aqueous solution
Catalog # DG097
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
:
:614.89 g/mol
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083
Melting Pt:116
Merck Index:13,02052