



Phenylmagnesium chloride 27% ∼2 mol/L in THF
Catalog # TCIAP1381-250G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):27.0 to 30.0 w/w%
Specification Test Results
About this item
:
:136.86 g/mol
Merck Index:14,07298
EINECS:202-868-8
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:100-59-4
Formula:C₆H₅ClMg
MDL Number:MFCD00000464