Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-Isopropylbenzyl chloride ≥85.0% (by GC) (contains ca. 10% o-form)
 :  TCI
 
undefined
 
4-Isopropylbenzyl chloride ≥85.0% (by GC) (contains ca. 10% o-form)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 85.0 %

 

Frequently Bought Together