Methyl-2,4-dimethoxybenzoate ≥98.0% (by GC)
Catalog # TCIAM2226-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:196.2 g/mol
Boiling Pt:296°C
CAS Number:2150-41-6
Formula:C₁₀H₁₂O₄
MDL Number:MFCD00070649