Order Entry
Netherlands
ContactUsLinkComponent
 
Corrosive
Irritant
Health
Environment
Boron trichloride ∼9% ∼1.0 mol/L in dichloromethane
Catalog # TCIAB3556-100ML
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
Health
Environment
Boron trichloride ∼9% ∼1.0 mol/L in dichloromethane
Catalog # TCIAB3556-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Neutralization titration):
    8.0 to 10.0 w/w%

Specification Test Results

About this item

 :
 :117.17 g/mol
Merck Index:14,01348
Storage Temperature:Refrigerator
EINECS:233-658-4
UN:3265
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
 :GHS09
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313