Methyl orange 0.1% in aqueous solution
Catalog # ROTHT119.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
About this item
:
:327.34 g/mol
Storage Temperature:Ambient
EINECS:208-925-3
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502