
2-(Tribromomethyl)quinoline ≥98.0% (by HPLC, titration analysis)
TCIAT1453-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:127.0 to 131.0 deg-C
:
:379.89 g/mol
:210-347-1
:GHS07
:613-53-6
:C₁₀H₆Br₃N
:MFCD00006755