Order Entry
Netherlands
ContactUsLinkComponent
(2-Methyl-3-nitrophenyl)acetic acid ≥96.0% (by GC, titration analysis)
Irritant
(2-Methyl-3-nitrophenyl)acetic acid ≥96.0% (by GC, titration analysis)
Catalog # TCIAM1817-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(2-Methyl-3-nitrophenyl)acetic acid ≥96.0% (by GC, titration analysis)
Catalog # TCIAM1817-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Neutralization titration):
    min. 96.0 %
  • Melting point:
    131.0 to 136.0 deg-C

Specification Test Results

About this item

 :
 :195.17 g/mol
 :GHS07
CAS Number:23876-15-5
Formula:C₉H₉NO₄
MDL Number:MFCD07782132