
2-Hydroxy-6-methylnicotinonitrile ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4241-27-4
:
Formula:C₇H₆N₂O
MDL Number:MFCD00006013
:134.14 g/mol
Melting Pt:295°C
EINECS:224-202-5
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 94.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together