
6,7-Dimethoxyisoquinoline ≥98.0% (by GC, titration analysis)
Catalog # TCIAD4089-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:92.0 to 96.0 deg-C
Specification Test Results
About this item
:
Boiling Pt:158°C
:GHS07
CAS Number:15248-39-2
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD00666134