
2-Methoxy-6-quinolineboronic acid pinacol ester 96%
Catalog # H54814.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:285.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1201644-36-1
Formula:C₁₆H₂₀BNO₃
MDL Number:MFCD18383956