Methyl-4-iodo-3-methylbenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5471-81-8
:
Formula:C₉H₉IO₂
MDL Number:MFCD02683863
:276.07 g/mol
Boiling Pt:106°C
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together