Methyl-3-methoxy-4-methylbenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3556-83-0
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00082710
:180.21 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together