
Probenecid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:57-66-9
:
Formula:C₁₃H₁₉NO₄S
MDL Number:MFCD00038402
:285.4 g/mol
Merck Index:14,07754
EINECS:200-344-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:198.0 to 202.0 deg-C
Specification Test Results
Frequently Bought Together