Order Entry
Netherlands
ContactUsLinkComponent
5-(Methylthio)-1H-tetrazole ≥98.0% (by HPLC)
Flammable
Irritant
5-(Methylthio)-1H-tetrazole ≥98.0% (by HPLC)
Catalog # TCIAM1889-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
5-(Methylthio)-1H-tetrazole ≥98.0% (by HPLC)
Catalog # TCIAM1889-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results

About this item

 :
UN:1325
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:29515-99-9
Formula:C₂H₄N₄S
MDL Number:MFCD00068726