
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2328-12-3
:
Formula:C₁₁H₁₆ClNO₂
MDL Number:MFCD00012744
:229.71 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together