5-Benzyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
TCIAB3086-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
:
:160.18 g/mol
:18489-25-3
:C₈H₈N₄
:MFCD00068730