


Di-tert-butyl pyrocarbonate ∼30% in THF
: TCI
:24424-99-5
:
:C₁₀H₁₈O₅
:MFCD00008805
:218.25 g/mol
:Refrigerator
:246-240-1
:1992
:3,II
:GHS02
:GHS06
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Morpholine Method):29.0 to 33.0 w/w%
Frequently Bought Together