Order Entry
Netherlands
ContactUsLinkComponent
Di-tert-butyl pyrocarbonate ~30% in 1,4-dioxane
Flammable
Toxic
Health
Di-tert-butyl pyrocarbonate ~30% in 1,4-dioxane
Supplier:  TCI
Certificates
undefined
Flammable
Toxic
Health
Di-tert-butyl pyrocarbonate ~30% in 1,4-dioxane
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Morpholine Method):
    29.0 to 33.0 w/w%

Specification Test Results

Frequently Bought Together