Order Entry
Netherlands
ContactUsLinkComponent
Cleanroom shirts with long sleeves, 7R014
Cleanroom shirts with long sleeves, 7R014
Supplier:  FRISTADS KANSAS
Certificates
undefined
Cleanroom shirts with long sleeves, 7R014
Supplier:  FRISTADS KANSAS
Specifications
  • Catalog No:
  • KWIN100640520L
  • KWIN100640520M
  • KWIN100640520S
  • KWIN100640520XL
  • KWIN100640520XS
  • KWIN1006405202XL
  • KWIN1006405203XL
  • Model:
  • 7R014, XA80
  • 7R014, XA80
  • 7R014, XA80
  • 7R014, XA80
  • 7R014, XA80
  • 7R014, XA80
  • 7R014, XA80
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • 100% filament polyester
  • 100% filament polyester
  • 100% filament polyester
  • 100% filament polyester
  • 100% filament polyester
  • 100% filament polyester
  • 100% filament polyester
  • Weight:
  • 99 g/m²
  • 99 g/m²
  • 99 g/m²
  • 99 g/m²
  • 99 g/m²
  • 99 g/m²
  • 99 g/m²