Order Entry
Netherlands
ContactUsLinkComponent
 
Toxic
Health
Formaldehyde 24%, VOLUSOL®, phosphate buffered (pH 7.0 ± 0.2)
Catalog # VWRS5322.9060
Supplier:  VWR Collection
CAS Number:  
Toxic
Health
Formaldehyde 24%, VOLUSOL®, phosphate buffered (pH 7.0 ± 0.2)
Catalog # VWRS5322.9060
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    60 L
  • Seq:
    5407

Specifications

About this item

 :
 :30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:3334
ADR:9,
 :GHS06
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274