Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Test/Sample Tube Closures
Screw caps for microcentrifuge tubes, 2,0 ml, conical
Screw caps for microcentrifuge tubes, 2,0 ml, conical
Screw caps for microcentrifuge tubes, 2,0 ml, conical
Catalog # BPLS4215R
Supplier:
BIO PLAS
Expand image
Screw caps for microcentrifuge tubes, 2,0 ml, conical
Catalog # BPLS4215R
Supplier:
BIO PLAS
Specifications
For:
Microcentrifuge tubes, with O-ring seal
Colour:
Natural
Pk:
1.000
Specifications
About this item
Product Details
Product Details